Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173546.2 | NP_775817.1 | 6 | 1485 | ||
XM_005264938.2 | XP_005264995.1 | 6 | 1971 | |||
XM_005264940.4 | XP_005264997.1 | 6 | 1606 | |||
XM_006713015.2 | XP_006713078.1 | 6 | 2001 | |||
XM_006713016.2 | XP_006713079.1 | 6 | 1980 | |||
GenBank mRNA | AK002164.1 | - | 2 | 1261 | ||
AK294160.1 | - | 6 | 1334 | |||
AY129011.1 | - | 6 | 1442 | |||
BC013110.2 | - | 2 | 478 | |||
BC039083.1 | - | 6 | 1435 | 5167173 |