Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDNRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256283.1 | NP_001243212.1 | 3 | 630 | ||
NM_001957.3 | NP_001948.1 | 4 | 1120 | |||
GenBank mRNA | AF014826.1 | - | 2 | 310 | ||
AK304451.1 | - | 3 | 402 | |||
AK312812.1 | - | 4 | 940 | |||
AK315931.1 | - | 3 | 615 | |||
AY275462.1 | - | 3 | 590 | |||
BC022511.1 | - | 4 | 940 | 4812050 | ||
D90348.1 | - | 4 | 1070 | |||
L06622.1 | - | 3 | 596 | |||
S45956.1 | - | 4 | 811 | |||
S57498.1 | - | 4 | 1074 | |||
S63938.1 | - | 3 | 663 | |||
S67127.1 | - | 3 | 727 | |||
S81539.1 | - | 3 | 590 | |||
X61950.1 | - | 4 | 1074 |