Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000043.5 | NP_000034.1 | 9 | 2036 | ||
NM_001320619.1 | NP_001307548.1 | 8 | 1953 | |||
NM_152871.3 | NP_690610.1 | 8 | 1973 | |||
NM_152872.3 | NP_690611.1 | 8 | 2011 | |||
NR_028033.3 | 7 | 1864 | ||||
NR_028034.3 | 6 | 1726 | ||||
NR_028035.3 | 7 | 1789 | ||||
NR_028036.3 | 8 | 1927 | ||||
NR_135313.1 | 7 | 1844 | ||||
NR_135314.1 | 9 | 2027 | ||||
NR_135315.1 | 7 | 1780 | ||||
XM_006717819.3 | XP_006717882.1 | 10 | 6718 | |||
XM_011539764.2 | XP_011538066.1 | 10 | 2107 | |||
XM_011539765.2 | XP_011538067.1 | 9 | 1832 | |||
XM_011539766.2 | XP_011538068.1 | 10 | 1847 | |||
XM_011539767.2 | XP_011538069.1 | 9 | 1770 | |||
GenBank mRNA | AB209361.1 | - | 10 | 5353 | ||
BC012479.1 | - | 9 | 2028 | 4514272 | ||
M67454.1 | - | 9 | 1912 | |||
X63717.1 | - | 9 | 1938 |