Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLB1L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080407.2 | NP_001073876.2 | 14, 15 | 1796 | ||
XM_011542566.2 | XP_011540868.1 | 13, 14 | 1866 | |||
XM_011542567.2 | XP_011540869.1 | 13, 14 | 1866 | |||
XM_011542568.2 | XP_011540870.1 | 13, 14 | 1830 | |||
XM_011542569.2 | XP_011540871.1 | 9, 10 | 1455 | |||
XM_011542570.2 | XP_011540872.1 | 9, 10 | 1419 | |||
XM_011542571.2 | XP_011540873.1 | 13, 14 | 1866 | |||
XM_011542572.1 | XP_011540874.1 | 9, 10 | 1014 | |||
XM_011542573.1 | XP_011540875.1 | 7, 8 | 950 | |||
XM_011542574.2 | XP_011540876.1 | 7, 8 | 1073 | |||
XM_017017155.1 | XP_016872644.1 | 14, 15 | 2694 | |||
XM_017017156.1 | XP_016872645.1 | 10, 11 | 2319 | |||
GenBank mRNA | AK074712.1 | - | 5, 6 | 502 | ||
AK124514.1 | - | 1, 2 | 2794 | |||
BC131702.1 | - | 5, 6 | 492 |