Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173588.3 | NP_775859.3 | 4 | 525 | ||
XM_011520030.2 | XP_011518332.1 | 2 | 3429 | |||
XM_011520031.2 | XP_011518333.1 | 2 | 3429 | |||
XM_017017607.1 | XP_016873096.1 | 3 | 3320 | |||
XM_017017608.1 | XP_016873097.1 | 3 | 1343 | |||
GenBank mRNA | AK095113.1 | - | 4 | 525 | ||
BC115396.1 | - | 4 | 476 | |||
BC115397.1 | - | 4 | 476 | 40068579 | ||
DA338373.1 | - | 4 | 483 |