Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTOGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK075060.1 | - | 1 | 47 | ||
AK096852.1 | - | 7 | 722 | |||
BC101016.2 | - | 8 | 679 | |||
BC101017.2 | - | 7 | 593 | |||
BC101018.1 | - | 7 | 593 |