Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFCAB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144933.1 | NP_001138405.1 | 8 | 644 | ||
NM_173503.3 | NP_775774.1 | 6 | 495 | |||
XM_011524380.1 | XP_011522682.1 | 6 | 494 | |||
XM_011524381.1 | XP_011522683.1 | 6 | 462 | |||
GenBank mRNA | AK098684.1 | - | 6 | 496 | ||
BC101752.1 | - | 6 | 468 | 8069258 | ||
BC101778.1 | - | 6 | 468 | 8069284 | ||
EG327376.1 | - | 8 | 644 |