Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199148.1 | NP_001186077.1 | 9, 10 | 1134 | ||
NM_173512.2 | NP_775783.1 | 8, 9 | 1068 | |||
XM_005246350.4 | XP_005246407.1 | 10, 11 | 1287 | |||
XM_006712337.3 | XP_006712400.1 | 9, 10 | 1283 | |||
XM_011510737.2 | XP_011509039.1 | 9, 10 | 1379 | |||
XM_011510742.2 | XP_011509044.1 | 7, 8 | 1075 | |||
XM_017003455.1 | XP_016858944.1 | 10, 11 | 1523 | |||
XM_017003456.1 | XP_016858945.1 | 9, 10 | 1457 | |||
XM_017003457.1 | XP_016858946.1 | 9, 10 | 1448 | |||
XM_017003458.1 | XP_016858947.1 | 9, 10 | 1408 | |||
XM_017003459.1 | XP_016858948.1 | 8, 9 | 1146 | |||
XM_017003460.1 | XP_016858949.1 | 10, 11 | 1523 | |||
XM_017003461.1 | XP_016858950.1 | 10, 11 | 1523 | |||
XM_017003462.1 | XP_016858951.1 | 10, 11 | 1523 | |||
XM_017003463.1 | XP_016858952.1 | 9, 10 | 920 | |||
XM_017003464.1 | XP_016858953.1 | 6, 7 | 1132 | |||
XM_017003465.1 | XP_016858954.1 | 5, 6 | 660 | |||
GenBank mRNA | AK097141.1 | - | 8, 9 | 1068 | ||
AK293998.1 | - | 9, 10 | 1095 | |||
BC125157.1 | - | 11, 12 | 1061 | 40120878 |