Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNKSR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173515.2 | NP_775786.2 | 5 | 1044 | ||
XM_011535485.2 | XP_011533787.1 | 5 | 3527 | |||
GenBank mRNA | AK055911.1 | - | 5 | 1084 | ||
AK309680.1 | - | 5 | 650 | |||
AY328891.1 | - | 5 | 1044 | |||
AY328892.1 | - | 5 | 1044 | |||
AY328893.1 | - | 5 | 1044 | |||
AY328894.1 | - | 2 | 327 | |||
BC060761.1 | - | 5 | 966 | 5299236 |