Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCMDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136160.1 | NP_001129632.1 | 11 | 1550 | ||
NM_001136161.1 | NP_001129633.1 | 11 | 1585 | |||
NM_173518.4 | NP_775789.3 | 11 | 1585 | |||
XM_005251174.2 | XP_005251231.1 | 10 | 1447 | |||
XM_006716427.3 | XP_006716490.1 | 11 | 1523 | |||
XM_006716429.2 | XP_006716492.1 | 11 | 1463 | |||
XM_006716433.3 | XP_006716496.1 | 9 | 1265 | |||
XM_011517467.2 | XP_011515769.1 | 11 | 1544 | |||
XM_011517468.2 | XP_011515770.1 | 10 | 1418 | |||
XM_011517469.2 | XP_011515771.1 | 11 | 1544 | |||
XM_017013140.1 | XP_016868629.1 | 9 | 1184 | |||
GenBank mRNA | AK098558.1 | - | 7 | 1122 | ||
AK302169.1 | - | 11 | 1550 | |||
AK302672.1 | - | 11 | 1544 | |||
AK303003.1 | - | 9 | 1341 | |||
AK304170.1 | - | 10 | 1447 | |||
AL833526.1 | - | 2 | 251 | |||
BC034576.1 | - | 11 | 1585 | 4825975 | ||
BC063108.1 | - | 10 | 1463 | 4690709 |