Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080551.2 | NP_001074020.2 | 2 | 487 | ||
NM_173521.4 | NP_775792.4 | 4 | 494 | |||
NR_109816.1 | 6 | 696 | ||||
XM_006716974.2 | XP_006717037.1 | 6 | 680 | |||
XM_006716975.2 | XP_006717038.1 | 6 | 680 | |||
XM_011518302.2 | XP_011516604.1 | 4 | 2489 | |||
XM_011518303.1 | XP_011516605.1 | 4 | 461 | |||
XM_011518309.1 | XP_011516611.1 | 6 | 680 | |||
XM_017014341.1 | XP_016869830.1 | 6 | 680 | |||
GenBank mRNA | AK057341.1 | - | 6 | 631 | ||
AK126055.1 | - | 2 | 487 | |||
AL833535.1 | - | 4 | 488 | |||
BC112357.1 | - | 6 | 624 | |||
BC144680.1 | - | 6 | 620 | |||
DB088747.1 | - | 2 | 488 |