Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM71D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173526.3 | NP_775797.2 | 5, 6 | 1204 | ||
XM_011536500.1 | XP_011534802.1 | 5, 6 | 1168 | |||
XM_011536502.1 | XP_011534804.1 | 5, 6 | 1103 | |||
XM_011536503.2 | XP_011534805.1 | 4, 5 | 2145 | |||
GenBank mRNA | AK093449.1 | - | 5, 6 | 1168 | ||
BC050401.1 | - | 5, 6 | 1136 | 5745029 |