Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284194.1 | NP_001271123.1 | 3 | 1689 | ||
NM_173540.2 | NP_775811.2 | 3 | 1567 | |||
GenBank mRNA | BC036037.1 | - | 3 | 1567 | 5271548 | |
BC100994.1 | - | 3 | 1686 | 40005868 | ||
BC100995.1 | - | 3 | 1686 | 40005869 | ||
BC100996.1 | - | 3 | 1564 | 40005870 | ||
BC100997.1 | - | 3 | 1564 | 40005873 |