Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256124.1 | NP_001243053.1 | 5 | 2730 | ||
NM_173547.3 | NP_775818.2 | 6 | 2796 | |||
XM_006721760.3 | XP_006721823.1 | 6 | 2833 | |||
XM_006721761.3 | XP_006721824.1 | 5 | 2767 | |||
XM_017024344.1 | XP_016879833.1 | 4 | 2557 | |||
GenBank mRNA | BC013181.1 | - | 1 | 1668 | ||
BC021259.2 | - | 6 | 2764 | 4128330 | ||
BC073831.1 | - | 3 | 1993 | |||
BC098412.1 | - | 1508 |