Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000362.4 | NP_000353.1 | 5 | 2331 | ||
GenBank mRNA | AK298126.1 | - | 6 | 1151 | ||
BC014277.2 | - | 5 | 1388 | 3689386 | ||
D45917.1 | - | 1 | 545 | |||
U14394.1 | - | 5 | 1432 | |||
X80791.1 | - | 1 | 410 |