Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173558.3 | NP_775829.2 | 11 | 1391 | ||
XM_017010429.1 | XP_016865918.1 | 11 | 1383 | |||
GenBank mRNA | AK092732.1 | - | 10 | 1600 | ||
AK097981.1 | - | 7 | 1275 | |||
AK131079.1 | - | 6 | 1219 | |||
AK290419.1 | - | 12 | 1384 | |||
AK301156.1 | - | 9 | 1124 | |||
BC023645.2 | - | 11 | 1391 | 5087501 | ||
BC053655.1 | - | 12 | 1336 | 5755893 | ||
BX648164.1 | - | 11 | 1298 |