Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173595.3 | NP_775866.2 | 25, 26 | 2903 | ||
XM_011538197.2 | XP_011536499.1 | 24, 25 | 3005 | |||
XM_017019183.1 | XP_016874672.1 | 24, 25 | 3086 | |||
XM_017019184.1 | XP_016874673.1 | 25, 26 | 2935 | |||
GenBank mRNA | AB190990.1 | - | 25, 26 | 2920 | ||
AK125379.1 | - | 25, 26 | 2920 | |||
AK126893.1 | - | 18, 19 | 2063 | |||
BC042070.1 | - | 5, 6 | 529 |