Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEXDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173620.2 | NP_775891.2 | 5, 6 | 871 | ||
XM_005256346.3 | XP_005256403.1 | 4, 5 | 459 | |||
XM_005256347.4 | XP_005256404.1 | 5, 6 | 595 | |||
XM_006722280.3 | XP_006722343.1 | 4, 5 | 434 | |||
XM_011523560.2 | XP_011521862.1 | 3, 4 | 402 | |||
XM_011523561.2 | XP_011521863.1 | 4, 5 | 440 | |||
XM_017024484.1 | XP_016879973.1 | 5, 6 | 595 | |||
XM_017024485.1 | XP_016879974.1 | 4, 5 | 459 | |||
GenBank mRNA | AK074405.1 | - | 4, 5 | 434 | ||
AK295197.1 | - | 1, 2 | 31 | |||
AK316085.1 | - | 23 | ||||
BC018205.1 | - | 4, 5 | 466 | 3464710 | ||
BC035561.1 | - | 5, 6 | 552 | 5211583 | ||
FM204887.1 | - | 4, 5 | 455 |