Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WEE2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015392.1 | 4 | 385 | |||
GenBank mRNA | AK098171.1 | - | 4 | 385 | ||
BC101359.2 | - | 4 | 639 |