Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128228.2 | NP_001121700.2 | 1 | 1712 | ||
GenBank mRNA | AK074735.1 | - | 1 | 1106 | ||
BC023530.2 | - | 1 | 720 | |||
BC048807.1 | - | 1 | 416 | |||
BC071831.1 | - | 1 | 1042 | |||
BC098411.1 | - | 1 | 1150 | |||
BC111500.1 | - | 1 | 841 | 4992533 | ||
BC128601.1 | - | 1 | 857 | 40132958 | ||
BC143385.1 | - | 1 | 874 | 9051891 |