Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173600.2 | NP_775871.2 | 23697 | |||
GenBank mRNA | AK093065.1 | - | 12 | 595 | ||
AY236870.1 | - | 12 | 595 | |||
HM801842.1 | - | 23697 |