Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3orf33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308229.1 | NP_001295158.1 | 4 | 487 | ||
NM_173657.2 | NP_775928.1 | 5 | 614 | |||
XM_005247379.4 | XP_005247436.1 | 4 | 1459 | |||
XM_011512710.2 | XP_011511012.1 | 3 | 412 | |||
XM_011512711.2 | XP_011511013.1 | 4 | 591 | |||
GenBank mRNA | AF115515.1 | - | 4 | 450 | ||
AK055701.1 | - | 5 | 593 | |||
AK289890.1 | - | 4 | 487 | |||
AL832165.1 | - | 4 | 472 | |||
BC042198.1 | - | 4 | 449 | |||
BC064898.1 | - | 4 | 459 | 6008321 | ||
DA275323.1 | - | 4 | 487 |