Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173689.6 | NP_775960.4 | 13 | 4245 | ||
NR_104603.1 | 8 | 3265 | ||||
XM_005251934.2 | XP_005251991.1 | 8 | 3265 | |||
XM_011518556.2 | XP_011516858.1 | 13 | 4218 | |||
XM_011518557.2 | XP_011516859.1 | 13 | 4247 | |||
XM_011518558.2 | XP_011516860.1 | 14 | 4607 | |||
GenBank mRNA | AK095783.1 | - | 4 | 882 | ||
AK124819.1 | - | 1 | 316 | |||
AK126775.1 | - | 8 | 3262 |