Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP11C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010986.2 | NP_001010986.1 | 25, 26 | 3080 | ||
NM_173694.4 | NP_775965.2 | 25, 26 | 3080 | |||
XM_017029439.1 | XP_016884928.1 | 26, 27 | 3086 | |||
XM_017029440.1 | XP_016884929.1 | 26, 27 | 3757 | |||
XM_017029441.1 | XP_016884930.1 | 27, 28 | 3166 | |||
XM_017029442.1 | XP_016884931.1 | 26, 27 | 3307 | |||
XM_017029443.1 | XP_016884932.1 | 26, 27 | 3085 | |||
XM_017029444.1 | XP_016884933.1 | 25, 26 | 3697 | |||
XM_017029445.1 | XP_016884934.1 | 26, 27 | 3087 | |||
XM_017029446.1 | XP_016884935.1 | 25, 26 | 3017 | |||
XM_017029447.1 | XP_016884936.1 | 26, 27 | 3290 | |||
XM_017029448.1 | XP_016884937.1 | 26, 27 | 3422 | |||
XM_017029449.1 | XP_016884938.1 | 25, 26 | 3028 | |||
GenBank mRNA | AJ580093.1 | - | 25, 26 | 3082 | ||
AJ580094.1 | - | 25, 26 | 3082 | |||
AK091552.1 | - | 2, 3 | 173 | |||
AK125474.1 | - | 6, 7 | 654 | |||
AK125986.1 | - | 15, 16 | 2070 | |||
AK131262.1 | - | 10, 11 | 1356 | |||
BC132728.1 | - | 25, 26 | 3018 | |||
BC132730.1 | - | 25, 26 | 3018 | |||
BX648320.1 | - | 12, 13 | 1522 | |||
BX648981.1 | - | 8, 9 | 1038 |