Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PAPD4 SIRNA Assays ›
Gene Symbol:
PAPD4
Gene Name:
PAP associated domain containing 4
Gene Aliases:
GLD2, TUT2
Chromosome Location:
Chr.5: 79612416 - 79686648 on Build GRCh38
UniGene ID:
Hs.418198
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
41252
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PAPD4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001114393.1
NP_001107865.1
4, 5
650
NM_001114394.1
NP_001107866.1
3, 4
925
NM_001297744.1
NP_001284673.1
4, 5
650
NM_001297745.1
NP_001284674.1
3, 4
925
NM_173797.3
NP_776158.2
4, 5
684
XM_011543219.2
XP_011541521.1
3, 4
929
XM_011543220.2
XP_011541522.1
4, 5
999
XM_011543221.2
XP_011541523.1
5, 6
1088
XM_011543222.2
XP_011541524.1
4, 5
1018
XM_011543223.2
XP_011541525.1
4, 5
638
XM_011543224.2
XP_011541526.1
4, 5
637
XM_011543225.2
XP_011541527.1
5, 6
599
XM_011543226.2
XP_011541528.1
5, 6
719
XM_011543227.2
XP_011541529.1
6, 7
808
XM_011543228.2
XP_011541530.1
3, 4
929
XM_011543229.2
XP_011541531.1
3, 4
929
XM_011543230.2
XP_011541532.1
3, 4
929
XM_011543231.2
XP_011541533.1
3, 4
929
XM_017009151.1
XP_016864640.1
5, 6
1076
XM_017009152.1
XP_016864641.1
4, 5
999
XM_017009153.1
XP_016864642.1
4, 5
999
XM_017009154.1
XP_016864643.1
5, 6
1076
XM_017009155.1
XP_016864644.1
4, 5
1018
XM_017009156.1
XP_016864645.1
5, 6
1308
XM_017009157.1
XP_016864646.1
3, 4
929
XM_017009158.1
XP_016864647.1
4, 5
999
XM_017009159.1
XP_016864648.1
4, 5
1018
XM_017009160.1
XP_016864649.1
5, 6
599
XM_017009161.1
XP_016864650.1
4, 5
1018
XM_017009162.1
XP_016864651.1
4, 5
606
GenBank mRNA
AK095818.1
-
4, 5
531
AL833136.1
-
3, 4
911
BC026061.1
-
5, 6
683
3640197
BC047581.1
-
4, 5
629
4824607
CN348312.1
-
4, 5
539
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotidyltransferase
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
hematopoietic progenitor cell differentiation
mRNA processing
RNA polyadenylation
histone mRNA catabolic process
polynucleotide adenylyltransferase activity
protein binding
ATP binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0