Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEXDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173620.2 | NP_775891.2 | 9, 10 | 1409 | ||
XM_005256346.3 | XP_005256403.1 | 8, 9 | 997 | |||
XM_005256347.4 | XP_005256404.1 | 9, 10 | 1133 | |||
XM_006722280.3 | XP_006722343.1 | 8, 9 | 972 | |||
XM_011523560.2 | XP_011521862.1 | 7, 8 | 940 | |||
XM_011523561.2 | XP_011521863.1 | 8, 9 | 978 | |||
XM_017024484.1 | XP_016879973.1 | 9, 10 | 1133 | |||
XM_017024485.1 | XP_016879974.1 | 8, 9 | 997 | |||
GenBank mRNA | AK074405.1 | - | 8, 9 | 972 | ||
AK295197.1 | - | 5, 6 | 1190 | |||
AK316085.1 | - | 4, 5 | 1182 | |||
BC018205.1 | - | 8, 9 | 1004 | 3464710 | ||
BC035561.1 | - | 9, 10 | 1090 | 5211583 | ||
FM204887.1 | - | 8, 9 | 993 |