Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PAPD4 SIRNA Assays ›
Gene Symbol:
PAPD4
Gene Name:
PAP associated domain containing 4
Gene Aliases:
GLD2, TUT2
Chromosome Location:
Chr.5: 79612416 - 79686648 on Build GRCh38
UniGene ID:
Hs.418198
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
41338
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PAPD4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001114393.1
NP_001107865.1
8
1110
NM_001114394.1
NP_001107866.1
7
1385
NM_001297745.1
NP_001284674.1
7
1385
NM_173797.3
NP_776158.2
8
1144
XM_011543219.2
XP_011541521.1
7
1389
XM_011543220.2
XP_011541522.1
8
1459
XM_011543221.2
XP_011541523.1
9
1548
XM_011543222.2
XP_011541524.1
8
1478
XM_011543223.2
XP_011541525.1
8
1098
XM_011543224.2
XP_011541526.1
8
1097
XM_011543225.2
XP_011541527.1
9
1059
XM_011543226.2
XP_011541528.1
9
1179
XM_011543227.2
XP_011541529.1
10
1268
XM_011543229.2
XP_011541531.1
7
1389
XM_011543230.2
XP_011541532.1
7
1389
XM_011543231.2
XP_011541533.1
7
1389
XM_011543232.2
XP_011541534.1
3
175
XM_017009151.1
XP_016864640.1
9
1536
XM_017009153.1
XP_016864642.1
8
1459
XM_017009154.1
XP_016864643.1
9
1536
XM_017009155.1
XP_016864644.1
8
1478
XM_017009156.1
XP_016864645.1
9
1768
XM_017009161.1
XP_016864650.1
9
1541
GenBank mRNA
AK095818.1
-
8
991
AL833136.1
-
7
1371
BC026061.1
-
9
1143
3640197
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001008372, XM_006231776, XM_008760639, BC085771.
This siRNA also targets the following transcripts in Mouse: NM_133905, BC016629, AK033141.
Panther Classification:
Molecular Function -
nucleotidyltransferase
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
hematopoietic progenitor cell differentiation
mRNA processing
RNA polyadenylation
histone mRNA catabolic process
polynucleotide adenylyltransferase activity
protein binding
ATP binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-97l6p:80/100.66.76.85:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline