Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM126B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321618.1 | NP_001308547.1 | 4 | 304 | ||
NM_001321619.1 | NP_001308548.1 | 5 | 515 | |||
NM_001321621.1 | NP_001308550.1 | 4 | 417 | |||
NM_001321622.1 | NP_001308551.1 | 6 | 633 | |||
NM_001321623.1 | NP_001308552.1 | 5 | 402 | |||
NM_001321624.1 | NP_001308553.1 | 4 | 304 | |||
NM_001321625.1 | NP_001308554.1 | 5 | 515 | |||
NM_001321626.1 | NP_001308555.1 | 5 | 535 | |||
NM_001321627.1 | NP_001308556.1 | 6 | 458 | |||
NM_001321628.1 | NP_001308557.1 | 6 | 571 | |||
NM_001321629.1 | NP_001308558.1 | 6 | 458 | |||
NM_173822.4 | NP_776183.1 | 5 | 402 | |||
XM_006712443.3 | XP_006712506.1 | 4 | 417 | |||
XM_011511002.2 | XP_011509304.1 | 5 | 432 | |||
XM_017003879.1 | XP_016859368.1 | 6 | 613 | |||
XM_017003880.1 | XP_016859369.1 | 5 | 1027 | |||
XM_017003881.1 | XP_016859370.1 | 5 | 463 | |||
XM_017003882.1 | XP_016859371.1 | 6 | 514 | |||
XM_017003883.1 | XP_016859372.1 | 6 | 857 | |||
GenBank mRNA | AK124184.1 | - | 5 | 400 | ||
AK315104.1 | - | 5 | 396 | |||
AL832234.1 | - | 4 | 261 | |||
AL832346.1 | - | 6 | 453 | |||
AL833403.1 | - | 5 | 386 | |||
BC039295.1 | - | 5 | 400 | 5267063 |