Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172811.1 | NP_001166282.1 | 6 | 703 | ||
NM_001172813.1 | NP_001166284.1 | 7 | 948 | |||
NM_001172814.1 | NP_001166285.1 | 5 | 790 | |||
NM_001172815.2 | NP_001166286.1 | 7 | 817 | |||
NM_173851.2 | NP_776250.2 | 4 | 760 | |||
GenBank mRNA | AK298294.1 | - | 8 | 951 | ||
AL713790.1 | - | 7 | 789 | |||
AY117411.1 | - | 4 | 530 | |||
BC126446.1 | - | 5 | 629 | |||
BC143953.1 | - | 5 | 630 | |||
EF560713.1 | - | 6 | 703 |