Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1AKMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318939.1 | NP_001305868.1 | 2 | 165 | ||
NM_174928.2 | NP_777588.1 | 2 | 229 | |||
NR_134934.1 | 2 | 165 | ||||
XM_017020432.1 | XP_016875921.1 | 2 | 264 | |||
GenBank mRNA | AK316604.1 | - | 2 | 163 | ||
AL597222.1 | - | 2 | 166 | |||
BC018091.1 | - | 2 | 189 | 4793121 | ||
DA514874.1 | - | 2 | 156 | |||
DQ823637.1 | - | 2 | 194 |