Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256113.1 | NP_001243042.1 | 1, 2 | 204 | ||
NM_175075.4 | NP_778250.2 | 2, 3 | 372 | |||
XM_006716232.3 | XP_006716295.1 | 1, 2 | 352 | |||
GenBank mRNA | AY194292.2 | - | 1, 2 | 190 | ||
BC136248.1 | - | 1, 2 | 351 | 9020235 | ||
BC136249.1 | - | 1, 2 | 351 | 9020236 | ||
CN430635.1 | - | 1, 2 | 183 | |||
DB461052.2 | - | 2, 3 | 366 | |||
FJ381685.1 | - | 1, 2 | 196 |