Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206955.1 | NP_001193884.1 | 19 | 2567 | ||
NM_001206956.1 | NP_001193885.1 | 11 | 1286 | |||
NM_175607.2 | NP_783200.1 | 20 | 2522 | |||
NM_175613.2 | NP_783302.1 | 11 | 1289 | |||
XM_006713004.3 | XP_006713067.1 | 19 | 2532 | |||
XM_011533425.2 | XP_011531727.1 | 19 | 2535 | |||
XM_011533426.2 | XP_011531728.1 | 20 | 2754 | |||
XM_011533427.2 | XP_011531729.1 | 20 | 2735 | |||
XM_011533428.2 | XP_011531730.1 | 18 | 2724 | |||
XM_011533429.2 | XP_011531731.1 | 18 | 2659 | |||
XM_011533430.2 | XP_011531732.1 | 18 | 2620 | |||
XM_011533431.2 | XP_011531733.1 | 17 | 5369 | |||
XM_017005782.1 | XP_016861271.1 | 20 | 2477 | |||
XM_017005783.1 | XP_016861272.1 | 21 | 3351 | |||
XM_017005784.1 | XP_016861273.1 | 19 | 2812 | |||
XM_017005785.1 | XP_016861274.1 | 20 | 2459 | |||
XM_017005786.1 | XP_016861275.1 | 18 | 2721 | |||
XM_017005787.1 | XP_016861276.1 | 18 | 2617 | |||
GenBank mRNA | AF549455.1 | - | 17 | 2565 |