Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEGR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173808.2 | NP_776169.2 | 2 | 404 | ||
XM_011541200.2 | XP_011539502.1 | 2 | 660 | |||
XM_011541201.2 | XP_011539503.1 | 2 | 660 | |||
XM_017000961.1 | XP_016856450.1 | 2 | 660 | |||
GenBank mRNA | AK092307.1 | - | 2 | 311 | ||
AK295481.1 | - | 2 | 302 | |||
BC036771.1 | - | 2 | 326 | 5576828 | ||
BX538014.1 | - | 2 | 308 |