Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267774.1 | NP_001254703.1 | 3 | 499 | ||
NM_001267775.1 | NP_001254704.1 | 3 | 298 | |||
NM_001267776.1 | NP_001254705.1 | 2 | 262 | |||
NM_001267777.1 | NP_001254706.1 | 3 | 298 | |||
NM_001267778.1 | NP_001254707.1 | 2 | 262 | |||
NM_174887.3 | NP_777547.1 | 2 | 262 | |||
GenBank mRNA | AA490722.1 | - | 2 | 178 | ||
AK297769.1 | - | 3 | 465 | |||
AK309232.1 | - | 3 | 488 | |||
AL713094.1 | - | 3 | 500 | |||
AY224601.1 | - | 3 | 448 | |||
BC002640.2 | - | 2 | 242 | 3609552 | ||
BC038094.1 | - | 2 | 228 | 3907361 | ||
BE619475.1 | - | 3 | 234 | |||
BQ639919.1 | - | 3 | 248 |