Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128324.2 | NP_001121796.1 | 4 | 531 | ||
NM_001282905.1 | NP_001269834.1 | 5 | 981 | |||
NM_001282906.1 | NP_001269835.1 | 5 | 633 | |||
NM_174890.3 | NP_777550.2 | 4 | 879 | |||
NR_104256.1 | 4 | 531 | ||||
XM_005271842.2 | XP_005271899.1 | 3 | 404 | |||
XM_005271843.2 | XP_005271900.1 | 3 | 571 | |||
XM_017016932.1 | XP_016872421.1 | 4 | 623 | |||
XM_017016933.1 | XP_016872422.1 | 4 | 490 | |||
XM_017016934.1 | XP_016872423.1 | 4 | 543 | |||
XM_017016935.1 | XP_016872424.1 | 3 | 293 | |||
XM_017016940.1 | XP_016872429.1 | 5 | 1039 | |||
XM_017016941.1 | XP_016872430.1 | 5 | 582 | |||
XM_017016942.1 | XP_016872431.1 | 3 | 571 | |||
XM_017016943.1 | XP_016872432.1 | 3 | 847 | |||
XM_017016944.1 | XP_016872433.1 | 3 | 404 | |||
XM_017016946.1 | XP_016872435.1 | 4 | 490 | |||
GenBank mRNA | AF311324.1 | - | 4 | 495 | ||
AK090704.1 | - | 5 | 705 | |||
AK097924.1 | - | 4 | 498 | |||
AK292420.1 | - | 5 | 582 | |||
AK292469.1 | - | 4 | 480 | |||
AK314395.1 | - | 4 | 491 | |||
BC045587.1 | - | 4 | 531 | 5269483 |