Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174897.2 | NP_777557.1 | 3 | 212 | ||
XM_017027663.1 | XP_016883152.1 | 3 | 260 | |||
GenBank mRNA | AF465767.1 | - | 3 | 212 |