Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1AKMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318939.1 | NP_001305868.1 | 3, 4 | 285 | ||
NM_174928.2 | NP_777588.1 | 3, 4 | 349 | |||
XM_017020432.1 | XP_016875921.1 | 3, 4 | 384 | |||
GenBank mRNA | AK316604.1 | - | 3, 4 | 283 | ||
BC018091.1 | - | 3, 4 | 309 | 4793121 | ||
DQ823637.1 | - | 3, 4 | 314 |