Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHYHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100876.1 | NP_001094346.1 | 4, 5 | 1133 | ||
NM_001100877.1 | NP_001094347.1 | 2, 3 | 241 | |||
NM_174933.3 | NP_777593.2 | 4, 5 | 1133 | |||
GenBank mRNA | AK054994.1 | - | 5, 6 | 645 | ||
AK095000.1 | - | 4, 5 | 1133 | |||
BC009373.2 | - | 4, 5 | 620 | 4121964 | ||
BC051300.1 | - | 2, 3 | 213 | 4843267 | ||
BC053853.1 | - | 2, 3 | 202 | 5209351 |