Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCSK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174936.3 | NP_777596.2 | 4 | 913 | ||
NR_110451.1 | 2 | 210 | ||||
GenBank mRNA | AK075365.1 | - | 4 | 842 | ||
AK122717.1 | - | 5 | 598 | |||
AK124635.1 | - | 1 | 1372 | |||
AK293870.1 | - | 5 | 893 | |||
AK297473.1 | - | 2 | 210 | |||
EF692496.1 | - | 4 | 551 |