Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042463.2 | NP_001035928.2 | 5 | 763 | ||
NM_174940.3 | NP_777600.3 | 5 | 739 | |||
GenBank mRNA | BC008671.1 | - | 5 | 687 | 3854028 | |
CK819789.1 | - | 5 | 553 |