Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP2A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005173.3 | NP_005164.2 | 12 | 1705 | ||
NM_174953.2 | NP_777613.1 | 12 | 1705 | |||
NM_174954.2 | NP_777614.1 | 12 | 1705 | |||
NM_174955.2 | NP_777615.1 | 12 | 1705 | |||
NM_174956.2 | NP_777616.1 | 12 | 1705 | |||
NM_174957.2 | NP_777617.1 | 12 | 1705 | |||
NM_174958.2 | NP_777618.1 | 12 | 1705 | |||
XM_011523881.2 | XP_011522183.1 | 12 | 1705 | |||
XM_011523882.2 | XP_011522184.1 | 12 | 1705 | |||
XM_011523884.2 | XP_011522186.1 | 12 | 1705 | |||
XM_011523885.1 | XP_011522187.1 | 12 | 1705 | |||
XM_011523888.2 | XP_011522190.1 | 12 | 1705 | |||
XM_011523889.1 | XP_011522191.1 | 12 | 1705 | |||
XM_011523892.2 | XP_011522194.1 | 12 | 1705 | |||
XM_017024692.1 | XP_016880181.1 | 12 | 1705 | |||
XM_017024693.1 | XP_016880182.1 | 10 | 1331 | |||
GenBank mRNA | AF068220.1 | - | 12 | 1656 | ||
AK292716.1 | - | 12 | 1705 | |||
BC035729.1 | - | 12 | 1660 | 5723849 | ||
Z69881.1 | - | 12 | 1538 |