Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287529.1 | NP_001274458.1 | 6, 7 | 1181 | ||
NM_174983.4 | NP_778148.2 | 6, 7 | 1208 | |||
XM_006722647.3 | XP_006722710.1 | 6, 7 | 1215 | |||
XM_011527684.2 | XP_011525986.1 | 6, 7 | 1214 | |||
GenBank mRNA | AL556500.3 | - | 1, 2 | 98 | ||
BC036706.2 | - | 6, 7 | 1160 | 4623094 | ||
BC068439.1 | - | 6, 7 | 1197 | 5314853 |