Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195214.1 | NP_001182143.1 | 4 | 530 | ||
NM_173834.3 | NP_776195.2 | 5 | 659 | |||
XM_011530923.2 | XP_011529225.1 | 5 | 1067 | |||
XM_011530924.1 | XP_011529226.1 | 5 | 441 | |||
XM_017029450.1 | XP_016884939.1 | 5 | 441 | |||
XM_017029451.1 | XP_016884940.1 | 4 | 938 | |||
GenBank mRNA | AK303992.1 | - | 4 | 260 | ||
BC012469.1 | - | 5 | 397 | 4452699 |