Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTCAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168364.1 | NP_001161836.1 | 6 | 711 | ||
NM_001321325.1 | NP_001308254.1 | 6 | 711 | |||
NM_173853.3 | NP_776252.2 | 6 | 711 | |||
GenBank mRNA | AY157576.1 | - | 6 | 681 | ||
AY358993.1 | - | 5 | 769 | |||
BC101687.1 | - | 6 | 710 | 8069193 | ||
BC101689.1 | - | 6 | 711 | 8069195 | ||
BC143574.1 | - | 6 | 710 | 9052085 | ||
BM794111.1 | - | 2 | 130 |