Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267774.1 | NP_001254703.1 | 6 | 825 | ||
NM_001267775.1 | NP_001254704.1 | 6 | 624 | |||
NM_001267776.1 | NP_001254705.1 | 5 | 588 | |||
NM_001267777.1 | NP_001254706.1 | 6 | 548 | |||
NM_001267778.1 | NP_001254707.1 | 4 | 484 | |||
NM_174887.3 | NP_777547.1 | 6 | 706 | |||
GenBank mRNA | AA490722.1 | - | 4 | 400 | ||
AF070643.1 | - | 1 | 1196 | |||
AY224601.1 | - | 6 | 774 | |||
BC002640.2 | - | 6 | 686 | 3609552 | ||
BQ639919.1 | - | 6 | 498 |
Gene Symbol: TMEM97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CA441136.1 | - | 1 | 88 |