Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM9C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174901.5 | NP_777561.1 | 3 | 425 | ||
XM_005274460.2 | XP_005274517.1 | 2 | 166 | |||
XM_017029321.1 | XP_016884810.1 | 2 | 166 | |||
GenBank mRNA | AF494345.1 | - | 3 | 308 | ||
AK313776.1 | - | 3 | 262 | |||
BC127957.1 | - | 2 | 119 | 40014693 | ||
BX647580.1 | - | 3 | 230 | |||
BX648111.1 | - | 3 | 309 | |||
BX648919.1 | - | 3 | 310 |