Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDILT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174924.1 | NP_777584.1 | 7, 8 | 1134 | ||
XM_011545764.1 | XP_011544066.1 | 7, 8 | 1165 | |||
XM_011545765.1 | XP_011544067.1 | 7, 8 | 1165 | |||
XM_011545766.2 | XP_011544068.1 | 3, 4 | 319 | |||
GenBank mRNA | AK097476.1 | - | 7, 8 | 1160 | ||
BC042607.1 | - | 7, 8 | 1134 | 5169707 | ||
BC044936.1 | - | 7, 8 | 1134 | 5169707 |