Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCN4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142348.1 | NP_001135820.1 | 3 | 511 | ||
NM_001142349.1 | NP_001135821.1 | 4 | 850 | |||
NM_174934.3 | NP_777594.1 | 5 | 913 | |||
NR_024527.1 | 4 | 696 | ||||
GenBank mRNA | AK124886.1 | - | 3 | 305 | ||
AK314531.1 | - | 5 | 826 | |||
AY149967.1 | - | 5 | 824 | |||
BC035017.2 | - | 4 | 814 | 4817294 | ||
BX538328.1 | - | 2 | 211 | |||
DA109496.1 | - | 3 | 492 |