Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001551.2 | NP_001542.1 | 7 | 1206 | ||
XM_017029489.1 | XP_016884978.1 | 6 | 1407 | |||
GenBank mRNA | AK054596.1 | - | 6 | 1407 | ||
AK315660.1 | - | 6 | 1324 | |||
BC004137.2 | - | 7 | 1206 | 2962967 | ||
BT006736.1 | - | 6 | 908 | |||
CR456782.1 | - | 6 | 908 | |||
Y08915.1 | - | 6 | 916 |