Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141919.1 | NP_001135391.1 | 11 | 1424 | ||
NM_001141920.1 | NP_001135392.1 | 10 | 1382 | |||
NM_175569.2 | NP_780778.1 | 10 | 1379 | |||
XM_005274587.3 | XP_005274644.1 | 11 | 2355 | |||
XM_011545575.2 | XP_011543877.1 | 8 | 2173 | |||
XM_017029787.1 | XP_016885276.1 | 10 | 2327 | |||
GenBank mRNA | AF380356.1 | - | 10 | 1357 |